Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACADL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109332 Supplier Invitrogen™ Supplier No. PA5109332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: SH-SY5Y cell, human kidney tissue, F9.

The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family, which is a family of mitochondrial flavoenzymes involved in fatty acid and branched chain amino-acid metabolism. This protein is one of the four enzymes that catalyze the initial step of mitochondrial beta-oxidation of straight-chain fatty acid. Defects in this gene are the cause of long-chain acyl-CoA dehydrogenase (LCAD) deficiency, leading to nonketotic hypoglycemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACADL
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4
Gene ACADL
Gene Accession No. P28330
Gene Alias AA960361; ACAD4; ACADL; acetyl-Coenzyme A dehydrogenase, long-chain; ACOADA; Acyl Coenzyme A dehydrogenase, long chain; acyl-CoA dehydrogenase long chain; acyl-CoA dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long chain; acyl-Coenzyme A dehydrogenase, long-chain; AU018452; C79855; LCAD; LCAD long chain acyl-CoA dehydrogenase; LOC100052267; long-chain acyl-CoA dehydrogenase; long-chain specific acyl-CoA dehydrogenase, mitochondrial; uncharacterized protein LOC100052267; zgc:55656
Gene Symbols ACADL
Host Species Rabbit
Immunogen Recombinant protein within Human ACADL aa 218-430.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 33
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.