Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACADM Monoclonal Antibody (OTI10A5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA811764

Description
This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
ACADM | |
Monoclonal | |
Unconjugated | |
ACADM | |
ACAD1, MCAD, MCADH | |
Mouse | |
Affinity Chromatography | |
RUO | |
34 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
OTI10A5 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P11310 | |
ACADM | |
Full length human recombit protein of human ACADM produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction