Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACADM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582237 Supplier Invitrogen™ Supplier No. PA582237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AWELGLMNTH IPENCGGLGL GTFDACLISE ELAYGCTGVQ TAIEGNSLGQ MPIIIAGNDQ QKKKYLGRMT EEPLMCAYCV TEPGAGSDVA GIKTKAEKKG DEYIINGQKM WITNGGKANW YFLLARSDPD PKA Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

This gene encodes the medium-chain specific (C4 to C12 straight chain) acyl-Coenzyme A dehydrogenase. The homotetramer enzyme catalyzes the initial step of the mitochondrial fatty acid beta-oxidation pathway. Defects in this gene cause medium-chain acyl-CoA dehydrogenase deficiency, a disease characterized by hepatic dysfunction, fasting hypoglycemia, and encephalopathy, which can result in infantile death. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACADM
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACADM
Gene Accession No. P11310
Gene Alias ACAD1; Acadm; acyl-CoA dehydrogenase C-4 to C-12 straight chain; acyl-CoA dehydrogenase, C-4 to C-12 straight chain; Acyl-Coenzyme A dehydrogenase C-4 to C-12 straight-chain; acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight chain; Acyl-Coenzyme A dehydrogenase, C-4 to C-12 straight-chain; acyl-Coenzyme A dehydrogenase, medium chain; AU018656; FLJ18227; FLJ93013; FLJ99884; MCAD; MCADH; medium-chain acyl-CoA dehydrogenase; medium-chain specific acyl-CoA dehydrogenase, mitochondrial; RP4-682C21.1; testicular tissue protein Li 7
Gene Symbols ACADM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACADM. Recombinant protein control fragment (Product #RP-88961).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 34
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.