Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACADS Antibody (8B10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259496H
This item is not returnable.
View return policy
Description
ACADS Monoclonal antibody specifically detects ACADS in Human samples. It is validated for Western Blot, ELISASpecifications
ACADS | |
Monoclonal | |
HRP | |
ACAD3, acyl-CoA dehydrogenase, C-2 to C-3 short chain, acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain, Butyryl-CoA dehydrogenase, EC 1.3.99, EC 1.3.99.2, mitochondrial short-chain specific acyl-CoA dehydrogenase, SCADshort-chain acyl-CoA dehydrogenase, short-chain specific acyl-CoA dehydrogenase, mitochondrial, unsaturated acyl-CoA reductase | |
Recombinant human ACADS (25-412aa) purified from E. coli | |
0.1 mL | |
Signal Transduction | |
35 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
8B10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction