Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACADSB Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | ACADSB |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
ACADSB Polyclonal specifically detects ACADSB in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
ACADSB | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
36 | |
IgG | |
Affinity purified |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
2-MEBCAD, 2-methylbutyryl-coenzyme A dehydrogenase, ACAD7,2-methylbutyryl-CoA dehydrogenase, acyl-CoA dehydrogenase, short/branched chain, acyl-Coenzyme A dehydrogenase, short/branched chain, EC 1.3.99,2-methyl branched chain acyl-CoA dehydrogenase, EC 1.3.99.-, SBCADshort/branched chain specific acyl-CoA dehydrogenase, mitochondrial | |
Recombinant fusion protein containing a sequence corresponding to amino acids 133-432 of human ACADSB (NP_001600.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title