Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACADSB Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197833
Description
ACADSB Polyclonal specifically detects ACADSB in Mouse, Rat samples. It is validated for Western Blot.Specifications
ACADSB | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 0.5% BSA, 5mM EDTA, 30% Glycerol with 0.01% Thimerosal | |
2-MEBCAD, 2-methylbutyryl-coenzyme A dehydrogenase, ACAD7,2-methylbutyryl-CoA dehydrogenase, acyl-CoA dehydrogenase, short/branched chain, acyl-Coenzyme A dehydrogenase, short/branched chain, EC 1.3.99,2-methyl branched chain acyl-CoA dehydrogenase, EC 1.3.99.-, SBCADshort/branched chain specific acyl-CoA dehydrogenase, mitochondrial | |
Rabbit | |
Affinity Purified | |
RUO | |
36 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/mL | |
Western Blot 1 - 4 ug/mL | |
Q9DBL1 | |
ACADSB | |
Synthetic peptide surrounding amino acid 399 of mouse ACADSB | |
0.1 mg | |
Primary | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction