Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADVL Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986273
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6273 Supplier Bioss Supplier No. BS5018R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is targeted to the inner mitochondrial membrane where it catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. This acyl-Coenzyme A dehydrogenase is specific to long-chain and very-long-chain fatty acids. A deficiency in this gene product reduces myocardial fatty acid beta-oxidation and is associated with cardiomyopathy. Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Antigen ACADVL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ACADVL
Gene Accession No. P49748
Gene Alias ACAD6; Acadvl; acyl-CoA dehydrogenase, very long chain; acyl-Coenzyme A dehydrogenase, very long chain; LCACD; MVLCAD; Very long chain Acyl-Coa dehydrogenase; Very long-chain specific acyl-CoA dehydrogenase, mitochondrial; Vlcad; VLCAD very-long-chain acyl-CoA dehydrogenase
Gene Symbols ACADVL
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ACADVL.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 37
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.