Learn More
Invitrogen™ ACAP2 Polyclonal Antibody

Description
This antibody is predicted to react with canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Specifications
Specifications
| Antigen | ACAP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ACAP2 |
| Gene Accession No. | Q15057, Q5FVC7, Q6ZQK5 |
| Gene Alias | 4832442G16; 9530039J15Rik; ACAP2; Arf GAP with coiled coil, ANK repeat and PH domains 2; arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2; ArfGAP with coiled-coil, ankyrin repeat and PH domains 2; centaurin beta2; centaurin, beta 2; centaurin-beta-2; Centb2; CNT-B2; KIAA0041; mKIAA0041 |
| Gene Symbols | ACAP2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.