Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566166 Supplier Invitrogen™ Supplier No. PA566166
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRQAWVQAVQA SIASAYRESP DSCYSERLDR TASPSTSSID SATDTRERGV KGESVL Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

ACAP3 is GTPase-activating protein for the ADP ribosylation factor family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACAP3
Gene Accession No. Q96P50
Gene Alias ACAP3; Arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 3; ArfGAP with coiled-coil, ankyrin repeat and PH domains 3; centaurin; centaurin, beta 5; centaurin-beta-5; CENTB5; cnt-b5; KIAA1716; Kiaa1716-hp; mKIAA1716
Gene Symbols ACAP3
Host Species Rabbit
Immunogen Recombinant Human ACAP3. Recombinant protein control fragment (Product #RP-106849).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.