Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAP4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
| Antigen | ACAP4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224666SS
![]() |
Novus Biologicals
NBP224666SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224666
![]() |
Novus Biologicals
NBP224666 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
ACAP4 Polyclonal specifically detects ACAP4 in Human, Primate samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACAP4 | |
| Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| Q8TDY4 | |
| 55616 | |
| Within the range of amino acids 693-729 of human ASAP3 were used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ARF6 GTPase-activating protein, arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3, ArfGAP with SH3 domain, ankyrin repeat and PH domain 3, ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1, centaurin, beta 6, CENTB6, DDEFL1, development and differentiation enhancing factor-like 1, Development and differentiation-enhancing factor-like 1, FLJ20199, Protein up-regulated in liver cancer 1, UPLC1ACAP4, up-regulated in liver cancer 1 (UPLC1) | |
| ASAP3 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a and CRA _b ), rhesus monkey, dog, mouse, cow and chimpanzee, and 75% homologus in pufferfish. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title