Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT1 Antibody (2C5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP222565AF594

Catalog No. NP2226A94


Only null left
Add to Cart

Description

Description

ACAT1 Monoclonal specifically detects ACAT1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ACAT1
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
ACAT1
Recombinant human ACAT1 (34-427aa) purified from E. coli
0.1 mL
Primary
Human
IgG1 κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1500000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL
Mouse
Protein A purified
Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction
38.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.