Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT1 Antibody (2C5), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222565MFV610
Description
ACAT1 Monoclonal antibody specifically detects ACAT1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ACAT1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 2C5 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL | |
| Recombinant human ACAT1 (34-427aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction | |
| 38 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction