Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP146140
Description
ACAT1 Polyclonal specifically detects ACAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
ACAT1 | |
Polyclonal | |
Western Blot 0.01 - 0.03 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:2000 | |
P24752 | |
ACAT1 | |
Peptide with sequence C-DEEYKRVDFSKVPK corresponding to internal region according to NP_000010.1. | |
Affinity Purified | |
RUO | |
Primary | |
This antibody recognizes and epitope within the internal region. Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Dog and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL | |
Goat | |
45 kDa | |
0.1 mg | |
Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction | |
38 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction