Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT1 (CES1) Mouse anti-Human,Monkey, Unconjugated, Clone: 10E8, MilliporeSigma™

Catalog No. MABC11011MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC11011MI 100 μg
MABC11012MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC11011MI Supplier MilliporeSigma Supplier No. MABC1101100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACAT1 (CES1) Monoclonal antibody specifically detects ACAT1 (CES1) in Human,Monkey samples. It is validated for ELISA, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAT1 (CES1)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10E8
Conjugate Unconjugated
Gene Accession No. NP_001020365
Gene Alias Liver carboxylesterase 1;EC: 3.1.1.13;Acyl-coenzyme A:cholesterol acyltransferase;ACAT;Brain carboxylesterase hBr1;Carboxylesterase 1;CE-1;hCE-1;Cocaine carboxylesterase;Egasyn;HMSE;Monocyte/macrophage serine esterase;Retinyl ester hydrolase;REH;Serine esterase 1;Triacylglycerol hydrolase;TGH
Gene Symbols CES1, CES2, SES1
Host Species Mouse
Immunogen A linear peptide corresponding to 14 amino acids from the C-terminal region of human Liver Carboxyesterase 1 (ACAT1).
Molecular Weight of Antigen ∼62 kDa observed; 62.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 10E8 is a mouse monoclonal antibody that specifically detects human liver carboxyesterase 1 (ACAT1). It targets an epitope with in 14 amino acids from the C-terminal region. It is expected to react with all isoforms of ACAT1.
Target Species Human, Monkey
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.