Learn More
Invitrogen™ ACAT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592402
Description
Immunogen sequence: MAVLAALLRS GARSRSPLLR RLVQEIRYVE RSYVSKPTLK EVVIVSATRT PIGSFLGSLS LLPATKLGSI AIQGAIEKAG IPKEEVKEAY MGNVLQGGEG QAPTRQAVLG AGLPISTPCT TINKVCASGM KAIMMASQSL MCGHQ; Positive Samples: SW620, Raji, NCI-H460, HepG2, U-937, Mouse kidney, Mouse liver, Mouse testis, Mouse small intestine; Cellular Location: Mitochondrion.
ACAT-1 is a ~50 kDa protein that catalyzes the formation of intracellular cholesterol esters in various tissues, such as adult human liver, adrenal glands, macrophages, and kidneys.
Specifications
ACAT1 | |
Polyclonal | |
Unconjugated | |
Acat1 | |
6330585C21Rik; Acat; Acat1; acetoacetyl Coenzyme A thiolase; acetoacetyl-CoA thiolase; acetyl-Co A acetyltransferase 1 mitochondrial; acetyl-CoA acetyltransferase 1; acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; RATACAL; T2; testicular tissue protein Li 198; THIL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
110446, 25014, 38 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.82 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P17764, P24752, Q8QZT1 | |
Acat1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-145 of human ACAT1 (NP_0000101). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.