Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582304
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582304 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582304 Supplier Invitrogen™ Supplier No. PA582304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEQDAYAINS YTRSKAAWEA GKFGNEVIPV TVTVKGQPDV VVKEDEEYKR VDFSKVPKLK TVFQKENGTV TAANASTLND GAAALVLMTA DAAKRLNVTP LARIVAFADA AVEPIDFPIA PVYAASMV Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 81%.

ACAT-1 is a ~50 kDa protein that catalyzes the formation of intracellular cholesterol esters in various tissues, such as adult human liver, adrenal glands, macrophages, and kidneys.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Acat1
Gene Accession No. P24752
Gene Alias 6330585C21Rik; Acat; Acat1; acetoacetyl Coenzyme A thiolase; acetoacetyl-CoA thiolase; acetyl-Co A acetyltransferase 1 mitochondrial; acetyl-CoA acetyltransferase 1; acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; RATACAL; T2; testicular tissue protein Li 198; THIL
Gene Symbols Acat1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACAT1. Recombinant protein control fragment (Product #RP-102378).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 38
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.