Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT1 Polyclonal Antibody, Biotin

ACAT1 Polyclonal Antibody, Biotin
Supplier: Thermo Scientific PA5142522
This item is not returnable.
View return policy
Description
ACAT1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| ACAT1 | |
| Polyclonal | |
| Biotin | |
| Acat1 | |
| 6330585C21Rik; Acat; Acat1; acetoacetyl Coenzyme A thiolase; acetoacetyl-CoA thiolase; acetyl-Co A acetyltransferase 1 mitochondrial; acetyl-CoA acetyltransferase 1; acetyl-CoA acetyltransferase, mitochondrial; acetyl-Coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; RATACAL; T2; testicular tissue protein Li 198; THIL | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 110446, 25014, 38 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P17764, P24752, Q8QZT1 | |
| Acat1 | |
| Peptide with sequence C-DEEYKRVDFSKVPK (aa 253-266). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction