Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986274
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6274 Supplier Bioss Supplier No. BS5019R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACAT-1 is a ˜50 kDa protein that catalyzes the formation of intracellular cholesterol esters in various tissues, such as adult human liver, adrenal glands, macrophages, and kidneys.

Specifications

Antigen ACAT1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Acat1
Gene Accession No. P24752
Gene Alias 6330585C21Rik; ACAT; Acat1; acetoacetyl Coenzyme A thiolase; Acetoacetyl-CoA thiolase; acetyl-Co A acetyltransferase 1 mitochondrial; acetyl-CoA acetyltransferase 1; Acetyl-CoA acetyltransferase, mitochondrial; acetyl-coenzyme A acetyltransferase 1; MAT; mitochondrial acetoacetyl-CoA thiolase; RATACAL; T2; testicular tissue protein Li 198; THIL
Gene Symbols Acat1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ACAT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 38
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.