Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT2 Antibody (55D5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242614H
Description
ACAT2 Monoclonal specifically detects ACAT2 in Human samples. It is validated for Western Blot, ELISA.Specifications
ACAT2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ACAT2 | |
Recombinant human ACAT2 (1-397aa) purified from E. coli. | |
100 μL | |
Cholesterol Metabolism, Lipid and Metabolism | |
39 | |
Store at 4C in the dark. | |
IgG2b κ |
Western Blot, ELISA | |
55D5 | |
Western Blot, ELISA | |
acetoacetyl Coenzyme A thiolase, acetyl-CoA acetyltransferase 2, acetyl-CoA acetyltransferase, cytosolic, Acetyl-CoA transferase-like protein, acetyl-Coenzyme A acetyltransferase 2, acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase), ACTL, Cytosolic acetoacetyl-CoA thiolase, EC 2.3.1, EC 2.3.1.9 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction