Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT2 Antibody (OTI3E2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270068H
Description
ACAT2 Monoclonal specifically detects ACAT2 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ACAT2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ACAT2 | |
Full length recombinant protein of human ACAT2 (NP_005882) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI3E2 | |
Western Blot, Immunohistochemistry | |
acetoacetyl Coenzyme A thiolase, acetyl-CoA acetyltransferase 2, acetyl-CoA acetyltransferase, cytosolic, Acetyl-CoA transferase-like protein, acetyl-Coenzyme A acetyltransferase 2, acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase), ACTL, Cytosolic acetoacetyl-CoA thiolase, EC 2.3.1, EC 2.3.1.9 | |
Mouse | |
Affinity Purified | |
Cholesterol Metabolism, Lipid and Metabolism | |
39.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction