Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT2 Antibody (SR1630), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | ACAT2 |
---|---|
Clone | SR1630 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
ACAT2 Monoclonal antibody specifically detects ACAT2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
ACAT2 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
acetoacetyl Coenzyme A thiolase, acetyl-CoA acetyltransferase 2, acetyl-CoA acetyltransferase, cytosolic, Acetyl-CoA transferase-like protein, acetyl-Coenzyme A acetyltransferase 2, acetyl-Coenzyme A acetyltransferase 2 (acetoacetyl Coenzyme A thiolase), ACTL, Cytosolic acetoacetyl-CoA thiolase, EC 2.3.1, EC 2.3.1.9 | |
A synthesized peptide derived from human ACAT2 (Uniprot #: Q9BWD1) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1630 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cholesterol Metabolism, Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
39 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title