Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT2 Monoclonal Antibody (OTI5F7), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

$231.00 - $516.29

Specifications

Antigen ACAT2
Clone OTI5F7
Concentration 1 mg/mL
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-254-2450
SDP
Encompass
Encompass_Preferred
View Documents
OriGene
TA501219S
30 μL
Each for $231.00
Only null left
Add to Cart
 
50-167-2364
SDP
Encompass_Preferred
View Documents
OriGene
TA501219
100 μL
Each for $516.29
Only null left
Add to Cart
 
Description

Description

ACAT2 Monoclonal antibody specifically detects ACAT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

ACAT2
1 mg/mL
Unconjugated
Mouse
IgG2b
-20° C, Avoid Freeze/Thaw Cycles
OTI5F7
Monoclonal
Liquid
RUO
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.