Learn More
ACAT2 Mouse anti-Canine, Human, Rat, Clone: OTI1C5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501216

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The product of this gene is an enzyme involved in lipid metabolism, and it encodes cytosolic acetoacetyl-CoA thiolase. This gene shows complementary overlapping with the 3-prime region of the TCP1 gene in both mouse and human. These genes are encoded on opposite strands of DNA, as well as in opposite transcriptional orientation.Specifications
ACAT2 | |
Monoclonal | |
Unconjugated | |
ACAT2 | |
acetyl-CoA acetyltransferase 2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
308100, 39, 484063 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5XI22, Q9BWD1 | |
Acat2 | |
Full length human recombit protein of human ACAT2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.