Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACBD3 Antibody (OTI1G2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272148AF750
Description
ACBD3 Monoclonal specifically detects ACBD3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
ACBD3 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ACBD3 | |
Full length recombinant protein of human ACBD3 (NP_073572) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1G2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
acyl-CoA binding domain containing 3, Acyl-CoA-binding domain-containing protein 3, acyl-Coenzyme A binding domain containing 3, GCP60Golgi phosphoprotein 1, GOCAP1Peripheral benzodiazepine receptor-associated protein PAP7, golgi complex associated protein 1, 60kDa, Golgi complex-associated protein 1, Golgi resident protein GCP60, GOLPH1PBR- and PKA-associated protein 7, PAP7, PBR associated protein, peripherial benzodiazepine receptor associated protein, PKA (RIalpha)-associated protein | |
Mouse | |
Affinity Purified | |
Golgi Apparatus Markers | |
64746.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction