Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACBD3 Monoclonal Antibody (OTI3A1), TrueMAB™, OriGene
SDP

Catalog No. 502540085
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0085 30 μL
50-167-5440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0085 Supplier OriGene Supplier No. TA504836S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACBD3 Monoclonal antibody specifically detects ACBD3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ACBD3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DPH2
Gene Accession No. Q9BQC3
Gene Alias 60kDa; 8430407O11Rik; Acbd3; acyl-CoA binding domain containing 3; acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; D1Ertd10e; DAP; DMT1-associated protein; fa20g08; GCP60; Gocap1; golgi complex associated protein 1, 60 kDa; golgi complex associated protein 1, 60kDa; golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; neurotrophin receptor alike death domain protein; p75-like apoptosis-inducing death domain protein; Pap7; PBR- and PKA-associated protein 7; PBR associated protein; Peripheral benzodiazepine receptor-associated protein PAP7; peripherial benzodiazepine receptor associated protein; PKA (RIalpha)-associated protein; si:dkey-267p15.1; wu:fa20g08; zgc:66303
Host Species Mouse
Immunogen Full length human recombinant protein of human DPH2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1802
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.