Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACBD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553331 Supplier Invitrogen™ Supplier No. PA553331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYPGNYEQQQ ILIRQLQEQH YQQYMQQLYQ VQLAQQQAAL QKQQEVVVAG SSLPTSSKVN ATVPSNMMSV NGQAKTHTDS SEKELEPEAA EEALENGPKE SLPVIAAPSM WTRPQIKDFK EKIQQDADSV IT Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.

GOLPH1, also known as GCP60, was initially identified as a Golgi protein that can interact with the integral membrane protein giantin and is thought to be involved in the maintenance of the Golgi structure. GOLPH1 has also been shown to interact with other Golgi proteins such as Golgin-160, a Golgi protein that can be cleaved by caspases-2, -3, and -7, leading to the nuclear localization of Golgin-160. GOLPH1 interaction with the Golgin-160 fragments is stronger than that with the intact Golgin-160, with its interaction regulated by the oxidation state of Cys-463 within GOLPH1, suggesting that the nuclear localization of the caspase-cleaved Golgin-160 fragments is a highly coordinated event. GOLPH1 has also been found to interact with Numb, a cytosolic signaling protein that mediates asymetric cell division of neural progenitor cells to a daughter progenitor cell and a neuron, suggesting that Golgi fragmentation and reconstitution during the cell cycle differentially regulate Numb signaling through changes in GOLPH1 subcellular distribution and may couple cell fate with cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Acbd3
Gene Accession No. Q9H3P7
Gene Alias 60kDa; 8430407O11Rik; Acbd3; acyl-CoA binding domain containing 3; acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; D1Ertd10e; DAP; DMT1-associated protein; fa20g08; GCP60; Gocap1; golgi complex associated protein 1, 60 kDa; golgi complex associated protein 1, 60kDa; golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; neurotrophin receptor alike death domain protein; p75-like apoptosis-inducing death domain protein; Pap7; PBR- and PKA-associated protein 7; PBR associated protein; Peripheral benzodiazepine receptor-associated protein PAP7; peripherial benzodiazepine receptor associated protein; PKA (RIalpha)-associated protein; si:dkey-267p15.1; wu:fa20g08; zgc:66303
Gene Symbols Acbd3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACBD3. Recombinant protein control fragment (Product #RP-90777).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64746
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.