Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACBD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116686 Supplier Invitrogen™ Supplier No. PA5116686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, HepG2, Mouse testis, Rat testis Immunogen sequence: MAAVLNAERL EVSVDGLTLS PDPEERPGAE GAPLLPPPLP PPSPPGSGRG PGASGEQPEP GEAAAGGAAE EARRLEQRWG FGLEELYGLA LRFFKEKDGK AFHPTYEEKL KLVALHKQVL MGPYNPDTCP EVGFFDVLGN DRRREWAALG NMSKEDAMVE.

GOLPH1, also known as GCP60, was initially identified as a Golgi protein that can interact with the integral membrane protein giantin and is thought to be involved in the maintenance of the Golgi structure. GOLPH1 has also been shown to interact with other Golgi proteins such as Golgin-160, a Golgi protein that can be cleaved by caspases-2, -3, and -7, leading to the nuclear localization of Golgin-160. GOLPH1 interaction with the Golgin-160 fragments is stronger than that with the intact Golgin-160, with its interaction regulated by the oxidation state of Cys-463 within GOLPH1, suggesting that the nuclear localization of the caspase-cleaved Golgin-160 fragments is a highly coordinated event. GOLPH1 has also been found to interact with Numb, a cytosolic signaling protein that mediates asymetric cell division of neural progenitor cells to a daughter progenitor cell and a neuron, suggesting that Golgi fragmentation and reconstitution during the cell cycle differentially regulate Numb signaling through changes in GOLPH1 subcellular distribution and may couple cell fate with cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Acbd3
Gene Accession No. Q7TNY6, Q8BMP6, Q9H3P7
Gene Alias 60kDa; 8430407O11Rik; Acbd3; acyl-CoA binding domain containing 3; acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; D1Ertd10e; DAP; DMT1-associated protein; fa20g08; GCP60; Gocap1; golgi complex associated protein 1, 60 kDa; golgi complex associated protein 1, 60kDa; golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; neurotrophin receptor alike death domain protein; p75-like apoptosis-inducing death domain protein; Pap7; PBR- and PKA-associated protein 7; PBR associated protein; Peripheral benzodiazepine receptor-associated protein PAP7; peripherial benzodiazepine receptor associated protein; PKA (RIalpha)-associated protein; si:dkey-267p15.1; wu:fa20g08; zgc:66303
Gene Symbols Acbd3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human ACBD3 (NP_0735722).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170760, 289312, 64746
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.