Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACBD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543439 Supplier Invitrogen™ Supplier No. PA543439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NSLGKMSREE AMSAYITEMK LVAQKVIDTV PLGEVAEDMF GYFEPLYQVI Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACBD4
Gene Accession No. Q8NC06
Gene Alias 2010009P05Rik; 2010015A21Rik; Acbd4; acyl-CoA binding domain containing 4; acyl-CoA-binding domain-containing protein 4; acyl-Coenzyme A binding domain containing 4; AI849317; FLJ13322; FLJ90623; HMFT0700; similar to acyl-Coenzyme A binding domain containing 4; zgc:85611
Gene Symbols ACBD4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ACBD4 (aa 74-123).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79777
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.