Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACBD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562477 Supplier Invitrogen™ Supplier No. PA562477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAYITEMKLV AQKVIDTVPL GEVAEDMFGY FEPLYQVIPD MPRPPETFLR RVTGWKEQVV NGDVGAVSEP PCLPKEPA Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 87%.

ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. All family members contain the conserved acyl-Coenzyme A binding domain, which binds acyl-CoA thiol esters. They are thought to play roles in acyl-CoA dependent lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACBD4
Gene Accession No. Q8NC06
Gene Alias 2010009P05Rik; 2010015A21Rik; Acbd4; acyl-CoA binding domain containing 4; acyl-CoA-binding domain-containing protein 4; acyl-Coenzyme A binding domain containing 4; AI849317; FLJ13322; FLJ90623; HMFT0700; similar to acyl-Coenzyme A binding domain containing 4; zgc:85611
Gene Symbols ACBD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACBD4. Recombinant protein control fragment (Product #RP-101537).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.