Learn More
ACBD5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303296AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Acyl-CoA binding protein which acts as the peroxisome receptor for pexophagy but is dispensable for aggrephagy and nonselective autophagy. Binds medium- and long-chain acyl-CoA esters.Specifications
ACBD5 | |
Polyclonal | |
Unconjugated | |
ACBD5 | |
acyl-CoA-binding domain-containing protein 5, acyl-Coenzyme A binding domain containing 5, endozepine-related protein, membrane-associated diazepam binding inhibitor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91452 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q5T8D3 | |
ACBD5 | |
Between 325 and 375 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.