Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACBD6 Antibody (4A9D5E9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306614V
Description
ACBD6 Monoclonal antibody specifically detects ACBD6 in Human samples. It is validated for Western Blot, ELISA, ELISA, ELISASpecifications
ACBD6 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
4A9D5E9 | |
Western Blot, ELISA, Sandwich ELISA, Sandwich ELISA Detection | |
acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACBD6 (NP_115736.1; Met 1-Ala 282). | |
0.1 mL | |
Cancer, Cell Biology, Endocrinology, Signal Transduction | |
84320 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction