Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACBD6 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297177APC
Description
ACBD6 Polyclonal antibody specifically detects ACBD6 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ACBD6 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404 | |
Produced in rabbits immunized with E. coli-derived Human ACBD6 fragment. | |
0.1 mL | |
Cancer, Cell Biology, Endocrinology, Signal Transduction | |
84320 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction