Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACBD6 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299708MFV450
Description
ACBD6 Polyclonal antibody specifically detects ACBD6 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
ACBD6 | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation | |
acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404 | |
Produced in rabbits immunized with purified, recombinant Human ACBD6 (Accession#: NP_115736.1; Met 1-Ala 282). | |
0.1 mL | |
Cancer, Cell Biology, Endocrinology, Signal Transduction | |
84320 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction