Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACBD6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555786
Description
Immunogen sequence: EGRALLHWAC DRGHKELVTV LLQHRADINC QDNEGQTALH YASACEFLDI VELLLQSGAD PTLRDQDGCL PEEVTGCKTV SLVLQRHTTG KA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
ACBD6 binds long-chain acyl-coenzyme A molecules with a strong preference for unsaturated C18:1-CoA, lower affinity for unsaturated C20:4-CoA, and very weak affinity for saturated C16:0-CoA. Does not bind fatty acids.
Specifications
ACBD6 | |
Polyclonal | |
Unconjugated | |
ACBD6 | |
0610010G04Rik; 2610100E10Rik; Acbd6; acyl-CoA binding domain containing 6; acyl-CoA-binding domain-containing protein 6; acyl-Coenzyme A binding domain containing 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84320 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BR61 | |
ACBD6 | |
Recombinant protein corresponding to Human ACBD6. Recombinant protein control fragment (Product #RP-94231). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction