Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ACBD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101401 Supplier Invitrogen™ Supplier No. PA5101401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACBD6.

ACBD6 binds long-chain acyl-coenzyme A molecules with a strong preference for unsaturated C18:1-CoA, lower affinity for unsaturated C20:4-CoA, and very weak affinity for saturated C16:0-CoA. Does not bind fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACBD6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACBD6
Gene Accession No. Q5RJK8, Q9BR61, Q9D061
Gene Alias 0610010G04Rik; 2610100E10Rik; Acbd6; acyl-CoA binding domain containing 6; acyl-CoA-binding domain-containing protein 6; acyl-Coenzyme A binding domain containing 6
Gene Symbols ACBD6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACBD6(Accession Q9BR61), corresponding to amino acid residues P130-S180.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289125, 72482, 84320
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.