Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACBD7 Antibody (005), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290286AF647
Description
ACBD7 Monoclonal specifically detects ACBD7 in Human samples. It is validated for ELISA.Specifications
ACBD7 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
414149 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
5 | |
ELISA | |
acyl-CoA binding domain containing 7, acyl-CoA-binding domain-containing protein 7, acyl-Coenzyme A binding domain containing 7, bA455B2.2, FLJ38219, FLJ52263, MGC33893 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ACBD7 (Q8N6N7; Met1-Ile88). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction