Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACBP Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297012R
Description
ACBP Polyclonal specifically detects ACBP in Human samples. It is validated for ELISA.Specifications
ACBP | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 550 | |
acyl-Coenzyme A binding domain containing 1, acyl-Coenzyme A bindingprotein), CCK-RP, cholecystokinin-releasing peptide, trypsin-sensitive, diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein), Diazepam-binding inhibitor, endozepine, EP, GABA receptor modulator, MGC70414 | |
Produced in rabbits immunized with purified, recombinant ACBP (P07108-2; Met1-Ala87). | |
0.1 mL | |
Cancer | |
1622 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction