Learn More
Thermo Scientific™ AccelerOme™ Label-Free MS Sample Prep Kit, 1 x 36 Reactions PROMO
Click to view available options
:
Thermo Scientific™ AccelerOme™ Label-Free MS Sample Prep Kit
Description
- Kit contains pre-formulated buffers, MS-grade trypsin/Lys-C enzyme mix, and peptide clean up cartridges for the processing of samples from protein lysis and digestion through peptide clean-up
- The single-use components are conveniently provided in labeled trays for direct placement on the AccelerOme system
- A peptide retention time calibration standard is also included with the kit (shipped separately) that can be spiked into the sample before LC-MS analysis to assess system performance.
- Complete set of ready-to-use proteomic sample prep and clean-up reagents for easy loading onto instrument
- Reagents combined with AccelerOme ecosystem provide consistent, high-quality results with technical replicates with <5% CV
- Reagents verified on system using cells, plasma, proteins, and tissue samples for 1–100 μg samples
- Instrumentation and reagents perform the work enabling time away from tedious hands-on sample preparation
Specifications
Specifications
Product Type | Reagent Kit |
Product Line | Thermo Scientific™ |
Starting Material | Protein Samples |
Detection Method | UV Absorption |
Quantity | 1 x 36 Reactions |