Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ AccelerOme™ System Test Kit PROMO
Click to view available options
:
Thermo Scientific™ AccelerOme™ System Test Kit
Description
- Kit provides control reagents for installation and QC of the AccelerOme Automated Sample Prep System
- The AccelerOme System Test Kit is required for installation of the AccelerOme Automated Sample Prep System
- Ready-to-use solutions provided in a tray that is directly inserted into the instrument
- Buffer matrix and peptide quantitation calibration standard monitor liquid handling and UV spectrophotometer system performance
- Designed specifically to provide installation and QC support of the AccelerOme system
- It contains a matrix buffer and peptide quantitation standard to assess the systems liquid handling reproducibility and accuracy
- Additionally, the quantitation standard serves as a quality control standard for the on-board UV spectrophotometer
- The system test kit may also be used for troubleshooting and routine instrument calibration
Specifications
Specifications
Product Type | Reagent Kit |
Product Line | Thermo Scientific™ |
Starting Material | Protein Samples |
Detection Method | UV Absorption |
Quantity | 1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction