Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beckman Coulter Access Total T4 Calibrators

Catalog No. 21280139 Shop All Beckman Coulter Products
Change view
Click to view available options
:
Access Total T4 Calibrators
Catalog No.
21-280-139 Access Total T4 Calibrators
1 options
This item is not returnable. View return policy

Catalog No. 21-280-139

Supplier: Beckman Coulter 33805

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Access Total T4 assay offers exceptional clinical quality for thyroid testing.

  • Measures total amount of free and bound thyroxine in circulation
  • Integrity; suppresses inflammatory and allergic activities
  • Broad dynamic range
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes Thyroxine (T4 Total)
For Use With (Equipment) Beckman Coulter Analyzer
Type Calibrator
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.