Learn More
Fisherbrand™ Accessories for Fisherbrand™ Multi-Purpose Tube Rotator
Catalog No. p-6731005 Shop All Fisherbrand Products
Click to view available options
Holds:
1.5/2.0 mL Tubes
10-15 mL and 5-7 mL Tubes
14 mm dia. Culture Tubes
15 mL Tubes
18.5 mm dia. Culture Tubes
50 mL Tubes
Optional accessories to enhance the Fisherbrand™Multi-Purpose Tube Rotator
Multi-purpose tube rotator offers flexibility with multiple carousel options
  • Tube rotator
TRUSTED_SUSTAINABILITY
Specifications
Holds 1.5/2.0 mL Tubes
Product Type Tube Carousel
No. of Positions 50
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.