Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diamedix™ Accessories for MAGO™ Plus and MAGO 4S Automated Immunoassay System


Click to view available options
Type:
Custom Mode Containers
Glass Wash Bottle, 2 L
Interference Filter, 550 nm
Reagent Rack, 330
Replacement Cap for 2 L Wash Bottle
Waste Container, 4 L
Description
For use with MAGO 4S and MAGO Plus Automated Immunoassay Systems.
Specifications
Specifications
Type | Reagent Rack, 330 |
For Use With (Equipment) | MAGO™ Plus and MAGO™ 4S automated immunoassay system |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction