Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Accessories for Samplicity™ Filtration Systems

Catalog No. p-7232203
Click to view available options
Type:
Replacement Lid
Tube Set Assembly
Material:
Silicone
Thermoplastic Elastomer

To be used with the Samplicity™ G2 Filtration System.

Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description Nonsterile lid with doors for attachment of Millex filters/funnels to HPLC tubes
For Use With (Equipment) Samplicity G2 Filtration System with Millex Filters
Material Thermoplastic Elastomer
For Use With (Application) Analytical sample prep; High Performance Liquid Chromatography (HPLC)
Housing Material PVDF
Quantity 1 Ea.
Sufficient For Up to 8 samples
Type Replacement Lid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.