Learn More
Thermo Scientific™ Accessories for TSG Series Countertop Refrigerators
Catalog No. tsgof02
Click to view available options
Type:
Omnicell Lock Adapter
Pyxis Lock Adapter
Innovative refrigeration solutions for healthcare and research needs
Thermo Scientific™ TSG Series Countertop Refrigerators have several accessory options available. E-lock adapter kits are available for Pyxis™ and Omnicell™ systems, and a 8.5 in. rollerbase makes moving the TSG205SA from room-to-room easy and convenient.
  • OF02 is compatible with Omnicellock™ systems
  • PA02 is compatible with Pyxis™lock systems
  • RB02 is an 8.5" roller base for the TSG205SA
TRUSTED_SUSTAINABILITY
Specifications
Type Omnicell Lock Adapter
Style Tabletop
For Use With (Equipment) TSG Series Countertop Refrigerator
Access Security Omnicell Lock Adapter
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.