Learn More
Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Catalog No. 502723593
Click to view available options
Product Type:
1536-well Adapter for 10 mm Plate w/o Lid
384-well Adapter for Plate w/ Lid
384-well Adapter for Plate w/o Lid
6-48-well Adapter for Plate w/o Lid
96-well Adapter for Plate w/ Lid
Aspirate tube assembly for a dispenser
Liquid Protection Tray
Robotic plate tray adapter for plates without lid
Upgrade Kit for Luminometry and TRF
Upgrade kit for luminometry and AlphaScreen
Upgrade kit for luminometry, AlphaScreen and TRF
Upgrade kit for two dispensers
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
50-272-3593 Aspirate tube assembly for a dispenser
50-950-607 Robotic plate tray adapter for plates without lid
50-950-608 384-well Adapter for Plate w/o Lid
50-950-609 384-well Adapter for Plate w/ Lid
50-950-611 96-well Adapter for Plate w/ Lid
50-950-612 6-48-well Adapter for Plate w/o Lid
50-950-622 1536-well Adapter for 10 mm Plate w/o Lid
N16308S Liquid Protection Tray
FEVLUPLA06 Upgrade kit for luminometry and AlphaScreen
FEVLUPLT07 Upgrade Kit for Luminometry and TRF
FEVLUPLAT08 Upgrade kit for luminometry, AlphaScreen and TRF
FEVLUPD209 Upgrade kit for two dispensers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 50-272-3593 Supplier Thermo Scientific™ Supplier No. 2805690
Add to Cart
Add to Cart
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.
Varioskan LUX is a modular instrument, you can upgrade it with missing features later.
  • Installation of the upgrade kits must be carried out by trained and authorized service personnel.
  • For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
  • TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
  • Please contact your local representative to discuss about the instrument upgrade option
Plate adapters are used for adapting plates that are of different heights.
  • Adapters adjust the plates to the optimum height for measuring and dispensing.
  • Choose the adapter based on the plate format in use and whether the lid is needed or not.
There are two types of plate trays: universal and robotic.
  • The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
  • A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Aspirate tube assembly for a dispenser
For Use With (Equipment) Varioskan LUX Microplate Reader
Product Line Varioskan