Description
- Use charge value with pressure-fuge head
- Maintenance Kit includes sample reservoir, filtrate container and 'O' ring seals
Specifications
Specifications
| Disposable | Reusable |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Vivacell 250 |
| Sterility | Non-sterile |
| For Use With (Application) | Maintenance Kit |
| Includes | Sample Reservoir, Filtrate Container, O-ring Seals |
| Type | Accessory for Vivacell Model |