Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Accessories for Wellwash™ and Wellwash™ Versa Microplate Washers

Catalog No. 502723561
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Aerosol Cover
Bottle Stand
Bottle Stand, 2 x 2 Configuration
Storage Bottle
Storage Bottle Cap
Wash Bottle A
Wash Head (Blank)
For Use With (Equipment):
Wellwash Microplate Strip Washer
Wellwash Versa Microplate Strip Washer
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
50-272-3561 Aerosol Cover Wellwash Microplate Strip Washer
50-272-3563 Wash Head (Blank) Wellwash Versa Microplate Strip Washer
50-272-3562 Wash Bottle A Wellwash Microplate Strip Washer
50-272-3560 Bottle Stand, 2 x 2 Configuration Wellwash Versa Microplate Strip Washer
50-272-3559 Bottle Stand Wellwash Microplate Strip Washer
50-272-3558 Storage Bottle Wellwash Versa Microplate Strip Washer
50-272-3557 Storage Bottle Cap Wellwash Microplate Strip Washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 50-272-3561 Supplier Thermo Scientific™ Supplier No. N10815
Add to Cart
Add to Cart

Enhance your Wellwash and Wellwash Versa microplate washers with a range of accessories and replacement parts. Wellwash microplate washers enable reliable ELISA plate washing for 96- or 384-well plates with low residual wash volumes and user-friendly features for optimal convenience and safety.

These accessories and replacement parts are for use with the Wellwash and Wellwash Versa microplate washers. Some are compatible with both models, while others are specifically designed for either the Wellwash or Wellwash Versa microplate washer.

Wellwash microplate strip washers provide reliable and robust plate washing for ELISA applications with low residual wash volumes. Wellwash microplate washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease of use, convenience, and safety.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
Type Aerosol Cover
For Use With (Equipment) Wellwash Microplate Strip Washer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.