Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Acclaim™ AmG C18 HPLC Columns
GREENER_CHOICE

Catalog No. p-6551584
Change view
Click to view available options
Length (Metric):
10 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
03-341-966 150 mm 2.1 mm
03-341-968 150 mm 3 mm
03-341-970 150 mm 4.6 mm
03-341-967 10 mm 2.1 mm
03-341-969 10 mm 3 mm
03-341-971 10 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-341-966 Supplier Thermo Scientific™ Supplier No. 088753
Only null left
Add to Cart
Add to Cart

Meet today’s demands for aminoglycoside antibiotic analyses, including drug purity characterization & quantification. They have unique tolerance to harsh conditions.

Get a high-performance, reversed-phase columns for aminoglycoside analysis. The robust and rugged Thermo Scientific™ Acclaim™ AmG C18 HPLC column provides exceptional tolerance required for aminoglycosides ion-pairing chromatographic separations. These silica-based C18 columns are optimized for various aminoglycoside antibiotic analyses, including drug purity characterization and quantification, therapeutic drug monitoring, and residual control testing in different matrices. These columns are stable at low pH, high temperature, organic solvents, and aqueous mobile phases.

Highly Stable for Aminoglycoside Analyses

The Acclaim AmG C18 column is specifically designed for ion-pairing reversed-phase HPLC (IP-RPLC) analysis of aminoglycoside antibiotics using volatile perfluorinated carboxylic acids as the ion-pairing reagent, such as 100 mM trifluoroacetic acid (TFA). The Acclaim AmG C18 stationary phase is based on a proprietary C18 bonding technology that provides extremely high stability towards acidic conditions. This specialty column exhibits excellent selectivity and high resolution for the aminoglycoside antibiotics analysis. In addition, it is easy to use because simple aqueous mobile phase (e.g. 100 mM TFA) without organic solvent and pH adjusting buffer is sufficient in most cases.

  • Excellent selectivity for aminoglycosides
  • Superior tolerance towards acidic conditions
  • High efficiency and throughput
  • Ease of use

The column can be used with UV, CAD, ELSD detectors

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 150 mm
Max. Pressure 8000 psi (550 bar)
Particle Size 3 μm
pH 0.5 to 10
Pore Size 120 Å
Quantity Each
Temperature <80°C
USP Type L1
Stationary Phase C18
Product Line Acclaim
Packing Material Spherical, High Purity Silica
Column Type Aminoglycoside Analysis
Phase Reversed Phase
Green Features FSC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.