Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion Chromatography HPLC Columns
GREENER_CHOICE

Catalog No. DX082739 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
33 mm
150 mm
300 mm
Diameter (Metric):
4.6 mm
7.8 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
DX082739 33 mm 4.6 mm
DX079721 300 mm 7.8 mm
DX079722 150 mm 7.8 mm
DX079724 300 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. DX082739 Supplier Thermo Scientific™ Supplier No. 082739
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.

Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion HPLC Columns. These resin-based, high-performance, size-exclusion chromatography columns cover molecular weight range: 7μm, 1000Å. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.

The Acclaim SEC-1000 LC column is calibrated from 1,000 to 1,000,000 Daltons and can characterize high molecular weight polymers.

High-Performance Analysis of Water-Soluble Polymers
  • High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
  • Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
TRUSTED_SUSTAINABILITY

Specifications

Column Format Guard Column
Diameter (Metric) 4.6 mm
Length (Metric) 33 mm
Particle Shape Spherical
Particle Size 7 μm
pH 2 to 12
Pore Size 1000 Å
Quantity Each
Surface Area 1000 m²/g
Temperature 60°C
Stationary Phase Proprietary
Product Line Acclaim
Packing Material Resin
Column Type Size Exclusion
Endcapped Yes
Phase Size Exclusion
Technique Bio-LC
Green Features FSC
Product Type Guard Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.