Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACCN4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215241
Description
ACCN4 Polyclonal specifically detects ACCN4 in Human samples. It is validated for Western Blot.Specifications
ACCN4 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
Acid-sensing ion channel 4, amiloride-sensitive cation channel 4, pituitaryMGC17248, ASIC4MGC24860, BNAC4amiloride-sensitive cation channel 4, brain sodium channel 4 | |
Rabbit | |
70 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (96%), Rabbit (95%), Rat (96%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.42mg/mL | |
Western Blot 1:500-1:3000 | |
Q96FT7 | |
ASIC4 | |
Recombinant protein encompassing a sequence within the center region of human ACCN4. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
55515 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction